ITT Jabsco Positive Displacement Pump Pulse Dampeners
Lieferant: Avantor Fluid Handling
74021-04
MFLX74021-04EA 854 EUR
MFLX74021-04
ITT Jabsco Positive Displacement Pump Pulse Dampeners
Pumpen Druckluftpumpen
Create a pulse-free flow out of your positive displacement pump.
  • Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.
More Info: These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.