FMI High-Pressure Piston Pump Accessories
Lieferant: Avantor Fluid Handling
07115-97 74200-80 74200-82 74200-83
MFLX07117-87EA 113 EUR
MFLX07117-87 MFLX07115-97 MFLX74200-80 MFLX74200-82 MFLX74200-83
FMI High-Pressure Piston Pump Accessories
Pumpen Kolbenpumpen
Make the most of your Masterflex® piston pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Adapt your pump to any existing system using the wide range of tubing adapters and assemblies. Replace rollers to extend pump life, or add a pulse dampener and/or cartridge heater for different applicaitons.