Edwards Vacuum Solenoid Right-Angle Valves
Lieferant: Avantor Fluid Handling
79302-78
MFLX79302-78EA 1650 EUR
MFLX79302-78
Edwards Vacuum Solenoid Right-Angle Valves
Ventile Solenoidventile
Designed to maintain vacuum levels in your systems.
  • Easily control vacuum levels with these solenoid valves
Compact and lightweight valves offer a pressure range from 8 x 10⁻¹⁰ to 150 Torr.