Dow Corning® Pharma-65 Transfer Tubing
Lieferant: Avantor Fluid Handling
96107-09
MFLX96107-01EA 204 EUR
MFLX96107-01 MFLX96107-04 MFLX96107-05 MFLX96107-11 MFLX96107-00 MFLX96107-02 MFLX96107-09
Dow Corning® Pharma-65 Transfer Tubing
Schläuche Transferschlauch
Platinum-cured silicone tubing with a very low extractable profile.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
Certification: Elastomer is FDA compliant and meets USP Class VI requirements. Meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Tubing is manufactured according to Bulk Pharmaceutical (BP) and current GMP. Temperature range: –58 to 482 °F (–50 to 250 °C).