MBD

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole

M-Clarity™ Qualitätsstufe = MQ100

MHS128-4L
MHS128-4LEA 269 EUR
MHS128-4L
MBD
MBD

Fluorescent probe for hydrophobic (non-polar) regions in proteins and nucleoproteins.

Formel: C₁₄H₁₂N₄O₄
Molecular Weight: 300,274 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00042778
CAS-Nummer: 33984-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f44fd15d254866a8e2fb47f9b9e02069daf2103d&vid=936d51d4cf333fe9286bdbf76e09d844b341d288&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Orange to Red
Appearance (Form) Powder
Proton NMR spectrum Conforms to Structure
Carbon 54.3 - 57.7 %
Nitrogen 18.1 - 19.3 %
Purity (HPLC) ≥97.5 %
Solubility (Colour) Orange
Solubility (Turbidity)
50 mg/ml DMSO:CH3OH (1:1)
Clear

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR