Valve brush
Lieferant: MICRONOVA
BRWH-30
MICRBRWH-30EA 151 EUR
MICRBRWH-30
Valve brush
Bürsten
Valve brush with oval head and 76 cm handle.
  • Polyester bristles
  • Base resins used to produce blocks and bristles are made from FDA approved materials
  • BPA-free