Valve brush
Lieferant: MICRONOVA
BRWH-50
MICRBRWH-50EA 118 EUR
MICRBRWH-50
Valve brush
Bürsten
Round head brush features medium stiff polyester bristles for aggressive cleaning.
  • BPA-free
  • Autoclavable
Base resins used to produce block and bristles are made from FDA approved materials.