Touch-free soap dispenser, MicroDispenser™
Lieferant: MICRONOVA
ICDISP-4
MICRICDISP-4EA 243 EUR
MICRICDISP-4
Touch-free soap dispenser, MicroDispenser™
Seifen- und Flüssigseifenspender
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas.
  • Reduces cross-contamination and waste
  • Activated by an electronic eye
  • Delivers a consistent, premeasured dose of soap, lotion, or sanitiser into the palm
  • Gravity fed with no tubes or pumps
  • No-drip unit may be wall-mounted
Easy delivery system allows dispenser to be use for different soaps or lotions by changing spout. Can be used with 500 ml pump bottles.
Standards: Complies with OSHA, CDC, and ADA requirements.
Lieferumfang: Operates on four size AA batteries which are included.