Mop heads, dry
Lieferant: MICRONOVA
MZDM62-14
MICRMZDM62-14EA 34.6 EUR
MICRMZDM62-14
Mop heads, dry
Mopps
Mops absorb detergent quickly and may be used on floors, walls, and ceilings without a bucket or wringer.
  • May be used with disinfectants
Material is designed for applications in which excess moisture is a concern. A special fabric strip allows for fast-drying action. Mop heads are sturdy enough to be laundered and reused. Latex/rayon blend mop head provides absorbency like that of PVA.