HPLC columns, Cogent UDC-Cholesterol™
Lieferant: MICROSOLV
69069-05P 69069-05P-1 69069-05P-2 69069-10P 69069-10P-1 69069-10P-2 69069-15P 69069-15P-1 69069-15P-2 69069-25P 69069-HG5 69069-HG6 69069-P21-100 69069-P21-150 69069-P21-250 69069-P21-50 69069-P30-100 69069-P30-150 69069-P30-250 69069-P30-50 69069-SP100 69069-SP150 69069-SP250 69069-SP50 69269-05P-1 69269-05P-2 69269-10P-1 69269-10P-2 69269-15P-2
MICS69069-05PEA 933 EUR
MICS69069-05P MICS69069-05P-1 MICS69069-05P-2 MICS69069-10P MICS69069-10P-1 MICS69069-10P-2 MICS69069-15P MICS69069-15P-1 MICS69069-15P-2 MICS69069-25P MICS69069-HG5 MICS69069-HG6 MICS69069-P21-100 MICS69069-P21-150 MICS69069-P21-250 MICS69069-P21-50 MICS69069-P30-100 MICS69069-P30-150 MICS69069-P30-250 MICS69069-P30-50 MICS69069-SP100 MICS69069-SP150 MICS69069-SP250 MICS69069-SP50 MICS69269-05P-1 MICS69269-05P-2 MICS69269-10P-1 MICS69269-10P-2 MICS69269-15P-2
HPLC columns, Cogent UDC-Cholesterol™
Chromatographiesäulen
The many advanced features of the this column produces a very stable, efficient and unique HPLC column with unique selectivity for steroids and other compounds.
  • Not endcapped liquid crystal
  • Shape recognition
  • 'Steroids' column
  • TYPE-C™ Silica
The UDC-Cholesterol™, an eleven carbon link with cholesterol attached is bonded to the silica hydride surface with a direct silicon-carbon bond. This phase is more sensitive to heat and solvent composition than most phases due to the liquid crystal nature of the cholesterol moiety. These parameters can be used inMore Product Information