Autosampler vials, screw top, 9-425
Lieferant: MICROSOLV
9502S-PP-CLEAR
MICS9502S-PP-CLEAREA 43.6 EUR
MICS9502S-PP-CLEAR
Autosampler vials, screw top, 9-425
Vials
These vials are the most commonly used autosampler vials for HPLC or GC. They are made of virgin, medical grade PP.
  • Pharmaceutical quality
  • Compatible with all leading instruments