Glass Inserts for 96 Well Plates, Microsolv tech corp
Lieferant: MICROSOLV
95096-710
MICS95096-710EA 192 EUR
MICS95096-710
Glass Inserts for 96 Well Plates, Microsolv tech corp
Vials
Glass inserts for 96 well plates, 1 ml, replacements. These borosilicate glass inserts are 1 ml and are replacements for 95096-1CP or 95096-3CP.
Bestellinformation: Plates and mats are sold separately.