Autosampler vials
Lieferant: MICROSOLV
9532C-0-WCV
MICS9532C-0-WCVEA 314 EUR
MICS9532C-0-WCV
Autosampler vials
Vials
These vials with serum finish are made from Type 1, first hydrolytic N51A grade borosilicate glass.
  • Used in gas chromatography
  • Used for storage of samples
Standard crimp top style vials for HPLC or GC autosamplers.