UV-Vis wavelenth standard
Lieferant: MICROSOLV
PQA-HOX-01
MICSPQA-HOX-01EA 297 EUR
MICSPQA-HOX-01
UV-Vis wavelenth standard
Standards Farbreferenzen und Spektroskopie-Standards
Wavelength calibration standard solution for UV-Vis detectors is composed of 4% Holmium oxide in 10% Perchloric acid.