Detergents for glassware washers, ProCare Lab 10 AP
Lieferant: MIELE
10265640 10265620
MIEL10265640EA 97 EUR
MIEL10265640 MIEL10265620
Detergents for glassware washers, ProCare Lab 10 AP
Reinigungsmittel Waschmittel und Oberflächenreiniger
Liquid, alkaline detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Removes stubborn or dried-on residues
  • Free from tensides, phosphates, oxidants
ProCare Lab 10 AP is intended for general use in medical, biological, microbiological and chemical laboratories, food-processing, cosmetics and pharmaceutical industries as well as the mineral oil sector and other branches.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, and aluminium.