Dishwashing Detergent, ProCare Universal 77 A
Lieferant: MIELE
12456030
MIEL12456030EA 306 EUR
MIEL12456030
Dishwashing Detergent, ProCare Universal 77 A
Reinigungsmittel Waschmittel und Oberflächenreiniger
Machine cleaner, alkaline for removing persistent starch deposits.
  • Removal of grease, oil and protein
  • Reduction in odours
  • Good cleaning capability
  • Very gentle on materials
  • High percentage of dispersants
  • Intensive cleaning agent containing surfactants
  • Strong emulsifying and dispersing effect
  • Specially developed for Miele Professional machines
Ingredients:
Phosphate
Oxygen-based bleaching agent
Phosphonates
Polycarboxylates
Non-ionic surfactants
Standards: GHS/CLP compliant.