Dishwashing Detergent, ProCare Universal 81 C
Lieferant: MIELE
12456130
MIEL12456130EA 168 EUR
MIEL12456130
Dishwashing Detergent, ProCare Universal 81 C
Reinigungsmittel Waschmittel und Oberflächenreiniger
Machine descaler, acidic, for removing persistent limescale deposits.
  • Removal of silicate deposits and other acid-soluble impurities
  • Prolongs service life of heater elements
  • Very gentle on materials
  • Very good material compatibility
  • Specially developed for Miele Professional machines
Contains citric acid and does not contain phosphorus compounds.
Standards: GHS/CLP compliant.