Natriumchlorid dishwasher salt, ProCare Universal 61 für die Enthärtung von Wasser
Lieferant: MIELE
ProCare Universal 61 is a coarse salt for reactivation of built-in water softener.
It prevents scaling and protects wash loads. Washing with softened water enables better cleaning with less cleaning agent required, better rinsing and reduced spotting.
Formel:
NaCl |
MDL:
MFCD00003477 CAS-Nummer: 7647-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...