Detergents, acid neutraliser, neodisher® Z
Lieferant: MIELE
US483030
MIELUS483030EA 0 EUR
MIELUS483030
Detergents, acid neutraliser, neodisher® Z
Reinigungsmittel Waschmittel und Oberflächenreiniger
neodisher® Z is a acid neutralising agent for the automated reprocessing of laboratory glassware for sensitive water examinations, laboratories in phosphate industry and also for the automated reprocessing of medical utensils in hospitals.
  • Free of surfactants and phosphates
  • Based on organic acids
  • Used to remove acid-soluble residues in laboratories where residues of phosphates, nitrogen and surfactants would adulterate sensitive analyses
neodisher® Z cleans stainless steel instruments, utensils and laboratory glassware as well as acidic pre-cleaning of laboratory glassware.
Standards: Conforms to the European directive 93/42/EC, Annex I, concerning medical devices.
Bitte beachten For avoiding water stains the use of deionised water in the final rinse is recommended.