Amicon® Stirred Cell Manifold
Lieferant: Merck
6015
MILF6015EA 930 EUR
MILF6015
Amicon® Stirred Cell Manifold
Verteiler
This manifold is designed for use with Amicon Stirred Cells and reservoirs and allows for operation of multiple cells or reservoirs.
  • Chemically sanitizable
  • Material: Delrin plastic, Buna-N
For instant direction of gas pressure or liquid flow in multi-cell or multi-reservoir systems. Can pressurize up to 3 cells or reservoirs from one gas source or feed several cells from one reservoir. Several manifolds can be combined to operate more than 3 units in parallel.
With this manifold, a single feed line can be distributed to three individual outlets. Several manifolds may be grouped together to provide additional outlet streams.