Minicon® Static Concentrators
Lieferant: Merck
The Minicon® Static concentrators to concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis. The B15 unit has 8 cells and can hold up to 5 ml of sample. The CS15 has 10 cells and is suited for volumes up to 2,5 ml. The absorbent pulls solvent and salts through the ultrafilter, concentrating the sample.
- Static concentrator, requiring no accessories
- Absorbent pulls solvent and salts through ultrafilter, concentrating sample
- Concentration factor up to 100X
- Low hold-up volume
- Nominal Molecular Weight Limit (NMWL): 15 kDa
Concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis (e.g., Bence Jones proteins in urine).
Applications: Concentration of urine and cerebrospinal fluid for enrichment of protein analytes (e.g., Bence Jones Proteins in Urine)
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...