Amicon® Pro Purification System
Lieferant: Merck
ACS500024
MILFACS500024EA
195
EUR
MILFACS500024
Amicon® Pro Purification System
Protein-Aufbereitungs
Protein Purification Kits
The Amicon® Pro purification system - A centrifugal tool collapsing protein purifcation with concentration and buffer exchange.
- Minimum final concentrate volume: 15 µl
- Non sterile
- Cap: Polyethylene
- Frit: Polypropylene/polyethylene
- Colour code: Clear with pink cap
The Amicon® Pro purification system 24 pack includes 24 exchange devices and 24 collection tubes with cap.
Applications include: Protein purification, protein concentration, buffer exchange, protein enrichment, antibody labeling.
Ø×Length: 29,7×120 mm
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...