Woven Mesh Spacer
Lieferant: Merck
AP3212450
MILFAP3212450EA 232 EUR
MILFAP3212450
Woven Mesh Spacer
Abstandshalter
Woven mesh spacer, dacron, 124 mm are placed between filters in serial filtration to prevent the downstream screen filter from 'blinding' the upstream filter pores. They increase flow rate and throughput.
  • Hydrophilic, made up of Polyethylene Terephthalate (PET)
  • Avoid membrane damage during high pressure or fast flow conditions
  • Spacers enable you to combine multiple stages of filtration into a single process step, saving time
Cellulose support pads are used to reinforce filters in Millipore's monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.
Filter code: AP32
Filter colour: White
Filter type: Net filter
Applications:
Air monitoring, environmental monitoring, aerosol contamination monitoring