EZ-Fit™ Manifold
Lieferant: Merck
EZFITMIC03
MILFEZFITMIC03EA 0 EUR
MILFEZFITMIC03
EZ-Fit™ Manifold
Verteiler
EZ-Fit™ Manifold for universal laboratory filtration.
  • Unique design for easy cleaning and decontamination
  • Different filtration heads for both reusable and disposable filtration devices
  • Quick-fit connectors for the vacuum tubing
  • Low height for ease of use in laminar flow hoods
  • Each component can be removed quickly and easily without any additional tools
  • A check valve is integrated into the tubing connection to prevent liquid back-flow from the vacuum system
  • The 2-way valve prevents the user from accidentally touching the filtration head while opening or closing it
The new EZ-Fit™ Manifold design eliminates sources of contamination that can cause false positive results: all internal areas are accessible and can be cleaned efficiently and all components can be autoclaved.