EZ-Stream™ Vacuum Pump
Lieferant: Merck
EZSTREAM
MILFEZSTREAMEA 0 EUR
MILFEZSTREAM
EZ-Stream™ Vacuum Pump
Pumpen Vakuumpumpen
Simply connect the pump to your manifold or filtration apparatus and place the discard tubing to drain and you are ready to filter. There is no need to use additional containers to capture the waste liquid. This brings a significant time saving versus traditional vacuum/pressure pumps or in-house vacuum because the following steps are eliminated: disconnecting the waste containers, emptying liquid waste, cleaning/decontaminating, re-installing the waste containers.
  • Compact design
  • High vacuum performance
  • EZ-Stream™ a cover protects the pump mechanismo reduce the noise down to 60 dB
  • EZ-Stream™ pump has been qualified to be compatible with a range of surface decontaminants
  • The diaphragm is made of highly durable chemically resistant polytetrafluoroethylene (PTFE)
  • EZ-Stream™ pump can provide vacuum up to 600 to 700 mbar
The EZ-Stream™ pump is at home handling both liquids and gases making it also suitable for many additional Life Science Laboratory applications.
The EZ-Stream™ pump technology and the materials of construction provide reliable vacuum performance and make the pump maintenance-free.
Standards: ISO 8199, IP 64, Protection class III.
Lieferumfang: Supplied with pump, multilingual user guide and power supply.