Millipore® Immobilon™-Ny+ Transfer Membranes
Lieferant: Merck
INYC00010
MILFINYC00010EA 0 EUR
MILFINYC00010
Millipore® Immobilon™-Ny+ Transfer Membranes
Transfer-Membranen
Web-supported positively charged nylon transfer membrane for Western blotting with chemiluminescence, radioactive and fluorescent detection methods.
  • Plain/white filter
  • Hydrophilic
  • Ambient shipping
Chemiluminescent, fluorometric and radioactive.