Multiscreen® Filter Plates, 96-Well, Hydrophobic PVDF Membrane
Lieferant: Merck
MAIPNTR10 MAIPSWU10
MILFMAIPNTR10EA 352 EUR
MILFMAIPNTR10 MILFMAIPSWU10
Multiscreen® Filter Plates, 96-Well, Hydrophobic PVDF Membrane
Mikrotiterplatten Mikrofiltrationsplatten
MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.
  • High binding surface
  • Flat bottom wells
  • Hydrophobic lid
  • Working volume: 50 to 250 µl
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.