MultiScreen® Collection Plates, 96 Well
Lieferant: Merck
MATRNPS50
MILFMATRNPS50EA 495 EUR
MILFMATRNPS50
MultiScreen® Collection Plates, 96 Well
Mikrotiterplatten HTS-Mikrotiterplatten
These 96 well collection plates are suitable for mammalian cell culture and parallel artificial membrane permeation assay (PAMPA).
  • Transport receiver plate
  • Clear
  • Non sterile
Made of polystyrene