Multiscreen® Filter Plates, 96-Well, Hydrophilic PVDF Membrane

Lieferant: Merck
MSHVN4B10 MSHVN4B50
MILFMSHVN4B10EA 352 EUR
MILFMSHVN4B10 MILFMSHVN4B50
Multiscreen® Filter Plates, 96-Well, Hydrophilic PVDF Membrane
Mikrotiterplatten Mikrofiltrationsplatten
MultiScreen® HTS HV Filter Plate, 0,45 μm, opaque, non-sterile has been used in the maltose-binding protein (MBP)-E6 and E6 association protein (AP) binding assay.

  • High throughput screening
  • Rigid sidewalls
  • Barcode-labeling capability
  • Design that prevents cross-well contamination

L×W×H: 128×85,5×14,4 mm
Filtration area: 0.28 cm²
Binding type: Low binding surface

Standards: The American National Standards Institute (ANSI) and the Society of Biomolecular Screening (SBS) standards compliance.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR