Multiscreen® Filter Plates with Negatively Charged Phosphocellulose
Lieferant: Merck
MSPHN6B10
MILFMSPHN6B10EA 398 EUR
MILFMSPHN6B10
Multiscreen® Filter Plates with Negatively Charged Phosphocellulose
Mikrotiterplatten Mikrofiltrationsplatten
Enzyme assays are widely used in life science research and compound screening. Filter plates, by facilitating the separation of the products from the reactants in enzymatic reactions, enable reliable, highly sensitive, and automation-compatible assays. MultiScreen®HTS filter plates set the standard for filtration-based enzyme assays.
  • Saves time and sample, reduces solvent and radioactive wastes
  • Multiple configurations to match your application
  • Automation compatible
  • Superior vacuum filtration and filtrate collection for better washing and consistent data
  • Dimension: 1280×855×146 mm
  • Pore size: 0,65 µm
These versatile systems are widely used for kinase, phosphatase, protease and endonuclease assays, as well as second messenger assays such as for cAMP, cGMP, phosphodiesterase (PDE), Nitric Oxide (NO), Ca²⁺ and inositols. The filtration-based protocol produces specific, reliable results that are wellreferenced in literature.
Applications: Protein kinase assays with peptide substrates, other enzyme assays, second messenger assays.