MultiScreen Solvinert Filter Plates with PTFE Membrane, 96-well
Lieferant: Merck
MSRLN0450 MSRPN0410 MSRPN0450 MSRLN0410 MDRLN0410 MDRPNP410
MILFMSRLN0450EA 1780 EUR
MILFMSRLN0450 MILFMSRPN0410 MILFMSRPN0450 MILFMSRLN0410 MILFMDRLN0410 MILFMDRPNP410
MultiScreen Solvinert Filter Plates with PTFE Membrane, 96-well
Mikrotiterplatten Mikrofiltrationsplatten
MultiScreen HTS 96-well filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate.
  • PTFE membrane (hydrophilic or hydrophobic)
  • Flat bottom wells
  • Non sterile
  • Polyolefin copolymer
The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS Vacuum Manifold, or by centrifugation.
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters.