Millipore® Immobilon®-P NOW Bundle Hydrophobic PVDF Transfer Membrane
Lieferant: Merck
NWKITIP
MILFNWKITIPEA 1030 EUR
MILFNWKITIP
Millipore® Immobilon®-P NOW Bundle Hydrophobic PVDF Transfer Membrane
Transfer-Membranen
Hydrophobic immobilon-P PVDF transfer membrane for western blotting using chemiluminescence, chromogenic or radioactive detection methods.
  • Electrostatic, hydrophobic
  • Plain/white filter
  • Western blotting
  • 0,45 µm pore size
  • Comes in an 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (70×85 mm) and midi-blots (85×135 mm).
Compatible stains: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination and Sypro® ruby.