Scepter™ 3.0 Automated Cell Counter Kits
Lieferant: Merck
PHCC360KIT PHCC30000 PHCC360050 PHCC340050 PHCC360250 PHCC340250 PHCC340500 PHCC360500 PHCC340KIT
MILFPHCC360KITEA 4180 EUR
MILFPHCC360KIT MILFPHCC30000 MILFPHCC360050 MILFPHCC340050 MILFPHCC360250 MILFPHCC340250 MILFPHCC340500 MILFPHCC360500 MILFPHCC340KIT
Scepter™ 3.0 Automated Cell Counter Kits
Zellzähler und Durchflusszytometer Elektronische Zellzähler
The Scepter™ 3.0 handhled automated cell counter provides the unparalleled precision of Coulter impedance counting in a convenient, handheld format. Newest version of the Scepter™ cell counter has been updated with features that make it more convenient, easy-to-use, and accurate.
  • Coulter impedance method which avoids common artifacts arising from image-based cell counting methods
  • Does not rely on user technique or manual calculation for accuracy
  • Counts thousands of cells per measurement for maximum precision
  • Unique ergonomic design, convenient for
More Product Information