Multiscreen® SEQ₃₈₄ Filter Plates

Lieferant: Merck
S384SEQ10 S384SEQ50
MILFS384SEQ10EA 0 EUR
MILFS384SEQ10 MILFS384SEQ50
Multiscreen® SEQ₃₈₄ Filter Plates
Mikrotiterplatten Mikrofiltrationsplatten
The MultiScreen384-SEQ filter plate is suited for use with most liquid handling systems. The filter plate operates by vacuum filtration enabling top-loading and recovery of sample. Our plate is designed with the needs of the high throughput laboratory in mind, meets SBS standards and is ideally suited for 'walk away' automation.

  • Automation friendly: Design and protocol saves time, money and improves reproducibility
  • Efficient dye terminator & salt removal: Recovers highly purified sequencing products for long, accurate sequencing reads
  • Eliminates dry-down step: Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
  • Saves reagents: plates suitable for use with ¹/₁₆ sequencing reactions
  • Proprietary membrane
  • Color code: Clear
  • Automation compatible: No

Application: Research category, assays, general filtration and nucleic acid.

Verpackung: Does not include reagents.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR