Millex® Glass Fiber Syringe Filter
Lieferant: Merck
SLAP05010
MILFSLAP05010EA 86.7 EUR
MILFSLAP05010
Millex® Glass Fiber Syringe Filter
Filter Spritzenvorsatzfilter
A 50 mm diameter non-sterile syringe filter with a 2 µm pore size glass fiber membrane.