SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Lieferant: Merck
TISNAP2MM
MILFTISNAP2MMEA 0 EUR
MILFTISNAP2MM
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Analysatoren Protein-Analysatoren
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Bestellinformation: This system includes the SNAP i.d. 2.0™ base, a mini blot holding frame,a midi blot holding frame, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.