Ultrafree®-CL Centrifugal Filter Units with Microporous Hydrophilic PTFE Membrane
Lieferant: Merck
UFC40LG25
MILFUFC40LG25EA 211 EUR
MILFUFC40LG25
Ultrafree®-CL Centrifugal Filter Units with Microporous Hydrophilic PTFE Membrane
Filter Zentrifugaleinheiten
These centrifugal filter units can be used for a number of applications, including removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.
  • Pre-sterilised units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 15 ml tubes
Hydrophilic PTFE membranes and PP housing.