Millivac™ Mini Vacuum Pump
Lieferant: Merck
XF5423050
MILFXF5423050EA 1010 EUR
MILFXF5423050
Millivac™ Mini Vacuum Pump
Pumpen Vakuum-Membranpumpen
Millivac™ Mini vacuum pumps are compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laborabory applications. A patented diaphragm design has reduced the footprint of the pumps compared to conventional models.
  • Use Millivac Mini Pump to filter water and aqueous solutions
  • Neither pump requires lubrication and neither is explosion proof
  • Lubrication not required
  • Quiet, compact design conserves previous lab bench space
Applications: Millivac™ Mini Pump for Water and Aqueous Solutions.