Filter holders, Hydrosol™
Lieferant: Merck
XX2004720
MILFXX2004720EA 1710 EUR
MILFXX2004720
Filter holders, Hydrosol™
Filterhalter
This filter holder is suitable for particulate and biological contamination analysis via vacuum filtration.
  • Not autoclavable with filter in place
Lieferumfang: Includes grounding set for use with flammable liquids.