Naval Patch Test Kit
Lieferant: Merck
Patch Test Kit is a complete solution for field-based collection and analysis of hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water, and lubricating oils.
- Monitors particulate contamination rapidly in hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water and lubricating oils
- Filter color rating and particle assessment scales correspond to recognised standard contamination levels
- Detects significant changes in cleanliness through dependable and sensitive test results
- Comes with filters for 100 tests
It allows the detection of significant changes in cleanliness through dependable, sensitive, colorimetric-based analysis.
Features:
- Contains enough expendable materials for 100 tests
- Contained within a lightweight, easy-to-carry case
- Filter color rating and particle assessment scales correspond to recognized standard contamination levels
Lieferumfang: Kit contains: MILFPD1504700 PetriSlides for contamination analysis, MILFSMWP02500 MF-Millipore Membrane Filter, 5,0 µm, 25 mm, white, plain, 100, MILFSMWP04700 MF-Millipore Membrane Filter, 5,0 µm, 47 mm, white, plain, 100, MILFXX6200006P Filter forceps, blunt end, stainless steel, MILFXX6200035 Hand Vacuum Pump, stainless steel, MILFXX6300120 Patch Test Filter Holder Assembly, MILFXX6504704 Solvent Dispensing Bottle, MILFXX6504707 Swinnex 25 mm with back-pressure screen, MILFXX6504709 PVC Bottle, 120 ml, MILFXX6504710 Tubing for vacuum, MILFXX6504713 Naval Fluid Color Guide 1.