Milli-Q SQPAK™ Final Filter
Lieferant: Merck
SQPAKFINM1
MILLSQPAKFINM1EA 166 EUR
MILLSQPAKFINM1
Milli-Q SQPAK™ Final Filter
Filter Filterkartuschen
SQPAK™ Final filter ensures high-quality, bacteria- and particulate-free ultrapure (Type 1) water at the point of dispense. The purification technology includes a microfiltration filter that removes bacteria and particulates >0,22 µm.
  • Easy replacement by simply twisting
  • Replacement is recommended after 6 months
The SQPAK™ Final filter is useful for lab applications such as chemistry and microbiology.