Milli-Q SQPAK™ RO Cartridge
Lieferant: Merck
SQPAKPR0M1
MILLSQPAKPR0M1EA 551 EUR
MILLSQPAKPR0M1
Milli-Q SQPAK™ RO Cartridge
Filter Filterkartuschen
The SQPAK™ RO cartridge produces pure RO water at up to 42 L/h. This cartridge′s RO membrane efficiently removes up to 99% of water contaminants such as bacteria, ions, particulates and organics.
  • Efficiently removes up to 99% of ions, bacteria, particulates and organics from feed water
  • Easy to replace thanks to the Twist and Lock mechanism
  • Need for cartridge replacement is indicated by an orange, blinking pictogram light on the system
Pure label and green colour of the SQPAK™ RO cartridge is repeated on the production station interface, to inform about its water purification purpose and guide replacement.