[EN]PURIFICATION SYSTEM MILLI-Q SQ 200P 1 * 1 ST
Lieferant: Merck
Pure and Ultra Pure Water System, Milli-Q®, Milli-Q® SQ 200P purification system, ultrapure (type 1) water dispensing module from a pressurised purified water loop, Flow rate: Up to 1,6 L/min ultrapure water from handset dispenser, 141×220×503 mm, 6 kg
Neuer Artikel bei VWR!