[EN]PURIFICATION SYSTEM MILLI-Q SQ 240C 1 * 1 ST
Lieferant: Merck
Pure and Ultra Pure Water System, Milli-Q®, SQ 240C purification system, combined pure RO water production station and ultrapure (Type 1) water dispensing module with flexible dispensing, Water quality (Type): Type I (ultra pure water), Flow rate: Up to 1,6 L/min ultrapure water from handset dispenser
Up to 6 L/min pure RO water from SQ gravity valve, 409×220×503 mm; 526×220×629 mm (Including handset dispenser), 11 kg without SQ switch 3,5 L tank, Operating weight with full tank: 17 kg
Up to 6 L/min pure RO water from SQ gravity valve, 409×220×503 mm; 526×220×629 mm (Including handset dispenser), 11 kg without SQ switch 3,5 L tank, Operating weight with full tank: 17 kg