The Milli-Q® SQ 240CV water purification system delivers both pure reverse osmosis (RO) water and ultrapure (Type 1) water. It benefits from a fixed preset volume dispenser that records and repeats the dispensing volume of ultrapure water.
This Milli-Q® SQ 240CV water purification system is 1 of 6 preset configurations available for the Milli-Q® SQ 2Series family of systems. For this configuration, the RO production station and ultrapure dispensing module are present as one unit and cannot be separated. It fulfills all lab water requirements for both pure RO and Type 1 water.
Simple, manual handset dispenser allows any new or untrained user to deliver ultrapure water at a varible flow rate, from 1,6 L/min to drop by drop. Reliable 18,2 MΩ·cm resistivity ultrapure (Type 1) water quality is indicated by a coloured LED, and TOC is reduced by an integrated photo-oxidation UV lamp. Freshly dispensed ultrapure water is available from the attached ultrapure water dispensing module; When completed with additional autonomous Milli-Q® SQ 200 ultrapure dispensing modules, ultrapure water is available anywhere with an electrical outlet thanks to the mobile SQ Switch tank.
Bestellinformation: This catalog number does not include:
SQPAK™ cartridges and final filter,
SQ Switch 3,5 L tank,
SQ Gravity valve or power cord adapted to the local electrical network.