The Milli-Q® SR 240 water purification system delivers pure reverse osmosis (RO) water to a mobile SQ Switch 3,5 L tank. This tank is easily transferred to the Milli-Q® SQ 200 Purification System to produce fresh ultrapure water anywhere in the lab. Thanks to the mobile SQ Switch tank, this water system adapts to any lab configuration: the RO water production unit is placed next to a potable water source, while unlimited ultrapure dispensing modules can be positioned anywhere in the lab or building with an electrical outlet. This allows for a flexible installation that can be easily adapted and scaled according to changing lab requirements.
The Milli-Q® SR 240 production station is part of 1 of 6 preset configurations (Milli-Q® SQ 240) available for the Milli-Q® SQ 2Series, which provides full autonomy to users and is adaptable to lab requirements and evolutions.
Fast (as little as 6 min) refilling of the mobile SQ Switch tank with freshly produced pure RO water thanks to the high-flow RO cartridge (up to 42 L/h); RO water recovery loop saves 50% of the rejected water; High performance maintained by automatic rinsing of RO membrane.
Compact setup and self-guided installation manual and videos let you plug the system wherever is convenient for your lab activity and move it later if needed
Unlimited scalability with one pure RO water production station able to feed multiple autonomous Milli-Q® SQ 200 ultrapure dispensing modules.This supports lab activity expansion while limiting budget investment
Bestellinformation: This catalog number does not include:
SQPAK™ cartridge,
SQ Switch 3,5 L tank,
SQ Gravity valve or power cord adapted to the local electrical network,
Ultrapure dispensing module – Milli-Q® SQ 200 Purification System.