Amicon® Stirred Cell Selector Valve
Lieferant: Merck
6003-M
MILX6003-MEA 872 EUR
MILX6003-M
Amicon® Stirred Cell Selector Valve
Ventile Auswahlventile
Valve with sliding control for instant switching from concentration to diafiltration, or switching gas and liquid lines simultaneously. Simplifies operation and avoids the need for multiple T-fittings and valves.
  • Device Material: Acetal, Buna-N and nylon
  • Chemically sanitizable
  • Dimension: 75×20×80 mm
The selector valve provides a quick and simple way to switch the Amicon Stirred Cell between concentration and diafiltration modes without interrupting system operation.
Application: The selector valve provides a quick and simple way to switch the Amicon Stirred Cell between concentration and diafiltration modes without interrupting system operation.
Key Applications: Concentration and diafiltration.