Milligard® Membrane Filter Units, PES 1,2/0,8 μm, Opticap® T-Line 30 Capsule
Lieferant: Merck
KMP8G3TTT1
MILXKMP8G3TTT1EA 1760 EUR
MILXKMP8G3TTT1
Milligard® Membrane Filter Units, PES 1,2/0,8 μm, Opticap® T-Line 30 Capsule
Filter Membranfiltergeräte
Milligard® PES 1.2/0.8 μm filters are designed for reliable particulate removal from a broad range of fluid streams. When used upstream of sterilizing-grade filters or other unit operations, they improve processing efficiency. Milligard® PES filters are supported by the Emprove® Program which consolidates product specific testing, quality and regulatory information to simplify compliance needs.
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer to Opticap® Small Scale 300 Capsule, XL, XLT and T-line Capsules User Guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations
Fast flow and high throughputPredictable scalability from small-scale to production-size devices High thermal stability: compatible with steam-in-place and autoclave sterilisation methods Single-use, gamma-ready formatsCaustic stable