Scepter™ 2.0 Automated Cell Counter
Lieferant: Merck
PHCC20060
MILXPHCC20060EA 0 EUR
MILXPHCC20060
Scepter™ 2.0 Automated Cell Counter
Zellzähler und Durchflusszytometer Elektronische Zellzähler
The Scepter cell counter accurately count particles smaller than 6 µm in diameter.
  • Easy-to-use, portable format
  • Counts blood cells and other cells with small diameters with the highest precision
The 40 µm sensor can count samples concentrated up to 1500000 cells/ml, 60 µm sensor can count up to 500000 cells/ml.
Powerful software for complex, effortless cell analysis, to compare sample sets side by side using histogram overlay and multiparametric tables, save and create gating templates, generate reports, graphs and tables.