Spectrophotometer cuvette
Lieferant: MINOLTA BUSINESS
1870-716
MINO1870-716EA 0 EUR
MINO1870-716
Spectrophotometer cuvette
Küvetten Spektralphotometer-Küvetten
Rectangular disposal cell suitable for transmittance measurements of water based transparent liquids.
Recommended for applications with liquids of normal density.